OH30378543
Ohaus® Dissolved Oxygen Meter
Model ST400D
About This Item
Codice UNSPSC:
41115611
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Ohaus® 30378543
dissolved oxygen measuring range
0.00-20.0 mg/L (0 - 50 °C)
Batteria
4 AAA, 250 hr life
Descrizione generale
Easy-to-use and accurate with a simple calibration process, automatic temperature compensation and fast results.
Large backlit LCD screen provides easy to read results - even in low light environments. Ideal for secure field use with a meter stand, wrist strap and durable IP54 housing.
Applicazioni
Field and lab Dissolved Oxygen measurement
Caratteristiche e vantaggi
99 measurement memory, last calibration data recall, auto barometric pressure measurement, auto temperature compensation with manual salinity compensation
Note legali
Ohaus is a registered trademark of Ohaus Corp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)