MTCP4280CR
MTC™ Bio Pipette Carousel
to hold, 6 Pipettes
Sinonimo/i:
conical tube, pipette holder, pipette rack
About This Item
Codice UNSPSC:
41122801
NACRES:
NB.04
Prodotti consigliati
Materiali
acrylonitrile-butadiene-styrene (ABS)
Caratteristiche
to hold 6 Pipettes
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
MTC™ Bio P4280-CR
Categorie correlate
Descrizione generale
This pipette carousel rack is perfect for keeping your bench-top organized. Universal fitting for pipettes, stable, and features slots for 6 single channel pipettors. Keep your pipettes safe and in a convenient location with this great pipette stand. MTC’s pipette carousel accepts many popular brands of pipettes on the market today. The list includes, but is not limited to: Gilson Pipetman® (all models, old and new), Rainin® (Classic and LTS), Eppendorf Research® Plus and various models of Labnet™ and HTL™ variable volume pipettes.
Caratteristiche e vantaggi
- Accepts most popular pipettes
- Stable, sturdy steel base
- Rotates 360 degrees
- Six pipette capacity
Note legali
MTC is a trademark of MTC Bio
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
MTCP4280CR | 4065270481993 |
MTCP4280CR-1EA | 4065271096417 |