Documenti fondamentali
L6289
Sigma-Aldrich
Laser Tough-Spots®
diam. 1/2 in., orange
Sinonimo/i:
solvent resistant labels, temperature resistant labels
About This Item
Codice UNSPSC:
55121611
NACRES:
NB.35
Materiali
orange
polyolefin
Confezionamento
pack of 3840 ea (192 labels per sheet)
Produttore/marchio commerciale
Diversified Biotech SPOT-2300
Diametro
1/2 in.
Descrizione generale
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Note legali
Tough-Spots is a registered trademark of Diversified Biotech
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)