L5539
Laser Tough-Spots®
diam. 3/8 in., green
Sinonimo/i:
solvent resistant labels, temperature resistant labels
About This Item
Prodotti consigliati
Materiali
green
polyolefin
Confezionamento
pack of 3840 ea (192 labels per sheet)
Produttore/marchio commerciale
Diversified Biotech SPOT-1200
Diametro
3/8 in.
Categorie correlate
Descrizione generale
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Note legali
Tough-Spots is a registered trademark of Diversified Biotech