Documenti fondamentali
HS29025C
Microtube Rack
green
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.12
Materiali
green
polypropylene
Caratteristiche
to hold 80 tube(s)
Confezionamento
pack of 5 ea
Produttore/marchio commerciale
Heathrow Scientific HS29025C
Lungh. × Largh. × Alt.
225 mm × 67 mm × 28 mm
Categorie correlate
Descrizione generale
The 80-Well Microtube Rack is a popular fraction-collector-plate style rack.
Caratteristiche e vantaggi
  • Easily fill and transfer 80 tubes in compact arrangement (5x16 array)
  • Frosted labeling area on bottom of rack
  • Embossed Index Numbers for quick sample ID and retrieval
  • Made of polypropylene
  • Autoclavable
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)