Documenti fondamentali
HS120592
Premium Plus Slide Box
to hold, 100 slide(s), translucent purple acrylonitrile-butadiene-styrene (ABS)
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.16
Materiali
polypropylene
translucent purple acrylonitrile-butadiene-styrene (ABS)
Caratteristiche
to hold 100 slide(s)
Confezionamento
pack of HS120592
Produttore/marchio commerciale
Heathrow Scientific 120592
Lungh. × Largh. × Alt.
208.3 mm × 195.6 mm × 33 mm
Descrizione generale
The Premium Plus Slide Box′s modern design for daily use is stylish with a wave surface texture. The slide box is easy to hold,increasing safety of sample transport with an EVA foam lined bottom of the box helping to hold slides securely in placeand protect against damage. Its unique side storage can accommodate a pen, desiccant or other small items needed for working.
Caratteristiche e vantaggi
  • Made from ABS and Polypropylene makes the boxes robust and eliminates potential rusting of metal parts
  • Inventory card is removable and replaceable for trouble-free writing on flat surfaces.
  • Hinged lid smoothly opens 180° sitting flat on surfaces and prevents misplacement
  • EVA foam lines the bottom of the box helping to hold slides securely in place and protect against damage
  • Multiple labeling areas, a large area on the lid of the box with “project” and “date” molded in and the back of the index card can be used as the lid is transparent
  • Vibrant translucent colors are easy to identify and enable coding options
  • Stylish wave surface texture is easy to hold, increasing safety of sample transport
  • Stackable, molded feet allow secure horizontal stacking of multiple units for safe transport or storing
  • Flexible storage, the units can be stored vertically or horizontally helping to efficiently use space available
  • Unique side storage can accommodate a pen, desiccant or other small items needed for working
  • Holds 100 - 26 x 76 mm slides
  • Patent Pending
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
HS120592-5EA4065265819404