HS159890B
Storage rack for 50 and 100 place microscope slide boxes
Holds 15 x slide boxes, clear acrylic
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.12
Prodotti consigliati
Materiali
clear acrylic
Caratteristiche
Holds 15 x slide boxes
Produttore/marchio commerciale
Heathrow Scientific HS159890B
Categorie correlate
Descrizione generale
Create a fi le system for 50- and 100-place slide boxes (see above)—storage rack makes organization of samples easy.
- Clear acrylic construction allows the labels to be seen on the slide boxes
- Place boxes in rows or columns by color to create an easy coding system
- Bins: 15
- Dimensions: 711 x 171 x 228 mm
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
HS159890B | 4061835296491 |