HS120245
Wide mouth wash bottle
white PE-LD bottle, capacity 500 mL
About This Item
Codice UNSPSC:
41103209
NACRES:
NB.43
Prodotti consigliati
Materiali
polypropylene cap
white PE-LD bottle
Caratteristiche
label no label
vented
with cap
Confezionamento
pack of 6 ea
Produttore/marchio commerciale
Heathrow Scientific HS120245
Capacità
500 mL
Categorie correlate
Descrizione generale
Color-coded tops for easy identification
The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
- Wide bottle mouth makes filling easier
- Color-coding for user, application or location identification
- Bottle is made from LDPE, with a PP closure
- 500 mL bottle size,
- Supplied in packs of 6
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
HS120245 | 4061835296378 |