Documenti fondamentali
G2528
Glass strips
For thick section (Histology) knives
About This Item
Codice UNSPSC:
41100000
NACRES:
MA.01
Descrizione
Pre-washed and ready to use
Confezionamento
pkg of 12 strips
Misura
8 in. (203 mm) × 1 1/2 in. (38 mm) × 1/4 in. (6 mm)
Descrizione generale
Specially formulated for microtomy knife making.