Documenti fondamentali
EP951020600
Eppendorf® twin.tec PCR Plates 96
Optimized rim for barcodes, semi-skirted, well volume 250 μL, colorless, pkg of 300 ea
About This Item
Codice UNSPSC:
41106300
Materiali
colorless
conical bottom
polypropylene wells
Sterilità
non-sterile
Caratteristiche
lid: no
semi-skirted
Confezionamento
pkg of 300 ea
Misura
96 well
Volume pozzetto
250 μL
Compatibilità
suitable for PCR
Descrizione generale
Eppendorf® twin.tec PCR Plate 96, semi-skirted, 250 µL, PCR clean, colorless, 300 plates
  • Semi-skirted for label and barcode
  • Fits most thermal cyclers
  • 250 µL max. well volume (when used with cap strips)
  • Improved well-to-well tolerance
  • Available with barcode (upon request), for skirted and semi-skirted only
Caratteristiche e vantaggi
  • Semi-skirted for label and barcode
  • Fits most thermal cyclers
  • 250 µL max. well volume (when used with cap strips)
  • Improved well-to-well tolerance
  • Available with barcode (upon request), for skirted and semi-skirted only
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)