Key Documents
EP5804732006
Eppendorf® Adapter
holds drum rotor 0.4 mL, pack of 6 pieces
About This Item
Codice UNSPSC:
41103911
NACRES:
NB.82
Caratteristiche
holds drum rotor 0.4 mL
Confezionamento
pack of 6 pieces
Produttore/marchio commerciale
Eppendorf® 5804732006
tecniche
centrifugation: suitable
Compatibilità
for use with Rotor T-60-11
Note legali
Eppendorf is a registered trademark of Eppendorf AG