CLS6796420D
Corning® combo hotplate and stirrer with digital display
AC/DC input 230 V AC (Supplied with both 3-pin UK plug and 2-pin Euro plug), plate L × W 5 in. × 7 in., CE compliant
Sinonimo/i:
stirrers
About This Item
Codice UNSPSC:
41103800
NACRES:
NB.72
Prodotti consigliati
Caratteristiche
CE compliant
Confezionamento
case of 1
Produttore/marchio commerciale
Corning 6796-420D
Disponibilità
not available in USA and Canada
Parametri
5-550 °C temp. range
60-1150 rpm speed
Lungh. × largh. lastra
5 in. × 7 in.
Descrizione generale
- Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
- Microprocessor maintains consistent and repeatable temperature and stir speed settings.
- Bright LED HOT TOP icon lights up when top plate temperature is over 60 °C, even when heat control is turned off.
- Separate temperature sensor provides power cut off if unit overheats
- Extremely durable and heat resistant Pyroceram glass-ceramic top
- Small footprint and low profile
- Meets UL and cUL standards
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)