CLS6795610D
Corning® stirrer with digital display
AC/DC input 120 V AC, US 3-pin plug, plate L × W 10 in. × 10 in.
Sinonimo/i:
magnetic stirrer, magnetic stirrers, stirrers
About This Item
Codice UNSPSC:
41103803
NACRES:
NB.73
Prodotti consigliati
Confezionamento
case of 1
Produttore/marchio commerciale
Corning 6795-610D
Disponibilità
not available in EU
Parametri
60-1150 rpm speed
Lungh. × largh. lastra
10 in. × 10 in.
Descrizione generale
- Digital LED stir speed display indicates when set speed has been reached
- Microprocessor maintains consistent and repeatable stir speed settings
- Extremely durable and heat resistant Pyroceram glass-ceramic top
- Small footprint and low profile
- Meets UL and cUL standards
Altre note
individual units are not routinely calibrated, rpm speeds are approximate only
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)