Documenti fondamentali
CLS4519104
Corning® ProCulture® spinner flask accessories
Gas Delivery and Venting Fitting with 1/8 in. and 1/4 in. Inlets for Vertical Sidearm Flasks
Sinonimo/i:
container accessory
About This Item
Codice UNSPSC:
41122104
NACRES:
NB.43
Produttore/marchio commerciale
Corning 4519-104
Descrizione generale
Side arm fittings used for a variety of functions in angled sidearm Spinner Flasks during cell culture. Sidearm fittings provide a convenient way of transferring media into or out of flasks aseptically. They can also be used to sparge the cell culture medium with gases to enhance cell production. Fittings comprise of a PET insert with a Viton O-ring and a polypropylene sealing cap & 316 stainless steel tubes.
Dual tubes extend to the bottom of the flask, and are held in place by Noryl nuts with integrated ferrules.
Combination tubes has two different length tubes, one extends to the bottom of the flask; the other is a shorter 6′ length. Both tubes are held in place by Noryl nuts with integrated ferrules.
Note legali
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)