Documenti fondamentali
CLS440140
Corning® boss head support clamp
Sinonimo/i:
lab equipment accessory
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.76
Materiali
aluminum
Produttore/marchio commerciale
Corning 440140
Lunghezza
58 mm
Categorie correlate
Descrizione generale
Corning offers a 2 in. long right angle black anodized aluminum Boss Head clamp that will accept up to 7/16 in. diameter support rods. It is designed to be used with the Corning Temperature probe support clamp.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
CLS440140-1EA4061831699296