Key Documents
CLS431411
Corning® glass fiber membrane filter discs
for the 250 mL funnel, diam. 49.5 mm
Sinonimo/i:
filter accessory
About This Item
Codice UNSPSC:
40161507
NACRES:
NB.24
product name
Corning® square glass fiber prefilter, for the 250 mL funnel, diam. 49.5 mm
Materiali
glass fiber membrane
Sterilità
non-sterile
Confezionamento
pkg of 100 ea
Produttore/marchio commerciale
Corning 431411
Diametro
49.5 mm
Note legali
Corning is a registered trademark of Corning, Inc.