CLS4082
Corning® Lambda® plus 8-channel pipettor
volume 20-200 μL
Sinonimo/i:
Multi-channel pipette
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Prodotti consigliati
Caratteristiche
8-channel
Produttore/marchio commerciale
Corning 4082
Volume
20-200 μL
Descrizione generale
- Corning® Lambda® plus 8-channel pipettor are designed for universal fit with all pipet tips of common brands.
- Adjustable tip ejector.
- For volume 20-200 μL.
- Thumb wheel or a plunger button to set volume
- Smooth plunger movement and extremely low pipetting forces.
- Accuracy (%) ±3.0 to ±1.0 and Precision (%) <1.5 to <0.6.
- 3-year warranty and Factory calibrated to strict ISO 8655 standards.
- Fully autoclavable and UV resistant.
- Inclusive of a key for easy in-house calibration.
Note legali
Corning is a registered trademark of Corning, Inc.
Lambda is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)