CLS3914
Corning® Medium Bind 1 x 8 Stripwell™ 96 well plates
96 well plate, 1 x 8 Stripwell 8-well strips, Medium Bind, polystyrene, flat bottom, black, non-sterile, 100/cs
Sinonimo/i:
1 x 8 strip plate, 8 well strip plate, 96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate
About This Item
Prodotti consigliati
Materiali
black polystyrene plate
flat bottom black wells
polystyrene plate
Sterilità
non-sterile
Caratteristiche
lid: no
skirt
plate format: 96 well Stripwell™ 8-well strips
Confezionamento
case of 100
pack of 25
Produttore/marchio commerciale
Corning 3914
Misura
96 wells
Volume pozzetto
360 μL
Volume di lavoro pozzetto
75-200 μL
Pozzetti
96 wells , stripwell
Compatibilità
suitable for (in vitro diagnostic assays)
Tipo di binding
non-treated surface
Descrizione generale
- Designed for in vitro diagnostic assays
- Flat bottom strips are designed to easily break apart and are pre-assembled in an "egg-crate" style strip holder that allows each individual well to be positioned back into the plate once broken
- Black wells have low background fluorescence, minimal light scatter and reduced crosstalk
- White wells enhance luminescent signals, have low background luminescence and fluorescence, and prevent well-to-well crosstalk
- Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
- Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
- Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm2
- Recommended working volume of 75 to 200 μL
- 1x8 Strips fit only one way into the strip holder, eliminating the chance of misorientation
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)