Documenti fondamentali
CLS3786
Corning® microcarriers
collagen coated, weight 10 g
Sinonimo/i:
lab equipment accessory
About This Item
Codice UNSPSC:
41122100
NACRES:
NB.13
Caratteristiche
collagen coated
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Corning 3786
Peso
10 g
Descrizione generale
These microcarriers are coated with Type I collagen to enhance cell attachment and expansion.
Corning Microcarriers are designed to simplify your scale-up processes. The microcarriers are sterile, ready to use, and available in closed systems packaging to be used directly with bioreactors.
Features:
  • USP Class VI polystyrene material provides a consistent platform
  • Level (SAL) of 10-6
  • Certified nonpyrogenic
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Lot/Batch Number
Ci dispiace, ma al momento non ci sono COA disponibili online per questo prodotto.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 3
1 of 3
Corning® microcarriers low concentration synthemax II, weight 100 g
CLS4622
Corning® microcarriers
Corning® microcarriers low concentration synthemax II, weight 500 g
CLS4623
Corning® microcarriers
Corning® microcarriers untreated, weight 500 g
CLS4625
Corning® microcarriers
Articoli
PLTMax Human Platelet Lysate (hPL) is a superior serum-free and xeno-free media supplement alternative to fetal bovine serum (FBS) for human mesenchymal stem cell (MSC) cultures.