CLS3004
Corning® single use spatulas
Small spoon/spoon
Sinonimo/i:
Spatula for Lab Use
About This Item
Codice UNSPSC:
41122403
NACRES:
NB.14
Prodotti consigliati
Sterilità
sterile
Produttore/marchio commerciale
Corning 3004
Lunghezza
236 mm
Descrizione generale
Corning single use spatulas have been designed to save time and provide contamination-free samples. Eliminates need to recycle and sterilize reusable spatulas.
Each spatula is individually packaged, RNase/DNase free, non-pyrogenic and antistatic.
Each spatula is individually packaged, RNase/DNase free, non-pyrogenic and antistatic.
Caratteristiche e vantaggi
- Corning® single use spatulas have a narrow spoon at one end and a larger round spoon at the other end.
- Polystyrene and color concentrate blend with 236 mm length.
- Visually Inspected for accurate labeling, product configuration, seal and barrier integrity.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)