Documenti fondamentali
BUA4389-EP
Buckets for Eppendorf® A-4-38 Rotor
pack of 4 ea, 90 mL rectangular
About This Item
Codice UNSPSC:
41103910
Caratteristiche
90 mL rectangular
Confezionamento
pack of 4 ea
Produttore/marchio commerciale
Eppendorf® 022639307
tecniche
centrifugation: suitable
Compatibilità
for use with Rotor A-4-38
Note legali
Eppendorf is a registered trademark of Eppendorf AG