Documenti fondamentali
AXYHS300
Axygen PlateMax heat sealing film for storage applications
pierceable, aluminum, 100 films/unit, 5 units/case
Sinonimo/i:
sealing mat
About This Item
Codice UNSPSC:
41122410
NACRES:
NB.13
Materiali
aluminum
Sterilità
non-sterile
Caratteristiche
piercable
Produttore/marchio commerciale
Axygen Inc HS-300
Corning
Descrizione generale
  • Axygen® PlateMax® heat sealing films are designed for applications ranging from PCR and Real-Time PCR to ELISA and cell culture.
  • Made from aluminum for use in wide temperature ranges.
  • Aluminum material provides a tight seal on multiwell microplates under a wide range of temperature conditions and reagents.
  • Seals during thermal cycling process when used in conjunction with Axygen compression matsIdeal for light-sensitive samples.
  • Compatible to use with the Axygen® PlateMax® Sem-Automated Plate Sealer (HS-1120, HS-1230).
Note legali
Axygen is a registered trademark of Corning, Inc.
PlateMax is a registered trademark of Axygen, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)