XXSPTFF01
Pellicon® Single-Pass TFF Mini Kit
About This Item
Codice UNSPSC:
23151800
eCl@ss:
32031690
Prodotti consigliati
tecniche
tangential flow filtration (TFF): suitable
Livello qualitativo
Descrizione generale
The Pellicon® Single-Pass TFF Mini Kit provides 4 diverter plates and 2 silicone gaskets to configure Pellicon® 3 88 cm2, Pellicon® 3 0.11 m² or Pellicon® 2 0.1 m2 cassettes for single-pass TFF operation.
Applicazioni
Increase product concentration and recovery. Run intensified, connected or continuous processes.
Note legali
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany