Documenti fondamentali
PA25/111
Millipore
Pneumatic Plastic Actuator for NOVASEPTIC® Valve, Normally Closed (NC)
About This Item
Codice UNSPSC:
31251504
Materiali
plastic cover (POM C)
plastic indicator (POM C)
stainless steel cover
Livello qualitativo
agenzia
meets EN ASTM
meets EN Stainless Steel EN 1.4305
Parametri
0-70 °C temp. range
1.5 kg pressure range (3.31 lb)
roughness average (RA)
1.6 μm ((63 μin) Electropolished)
Applicazioni
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Confezionamento
The Actuator is packaged in a closed box.
Note legali
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany