NA38/150
NovAseptic® Valve, Manual Plastic Actuator
About This Item
Codice UNSPSC:
40141600
eCl@ss:
37051001
Prodotti consigliati
Materiali
plastic handle (Oilon)
stainless steel cover (EN 1.4305/ASTM 303)
stainless steel indicator (SS 2346)
Livello qualitativo
Nome Commerciale
NovAseptic®
Parametri
pressure range at -1-10 bar
temp. range at 0-70 °C
Peso
1.2 kg (2.65 lb)
Applicazioni
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Altre note
Actuator, Manual
Note legali
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany