Z743198
Julabo® bath test tube racks
to fit 90 test tubes, diam. 11 mm (12 mm), stainless steel
Sinonimo/i:
tube holder
About This Item
Codice UNSPSC:
41103706
NACRES:
NB.77
Prodotti consigliati
Materiali
stainless steel
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Julabo 8970346
Parametri
100 °C max. temp.
Diametro
11 mm (12 mm)
Categorie correlate
Descrizione generale
Stainless steel rack suitable for temperatures up to 100°C. If solvent based samples or acidic samples are tempered above 60°C, it is recommended using test glass inserts made of stainless steel.
Note legali
Julabo is a registered trademark of Julabo USA, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)