Z742669
Artis Tweezer
Style MM-SA
About This Item
Codice UNSPSC:
27112105
NACRES:
MA.01
Prodotti consigliati
Caratteristiche
tip style (fine points.)
Produttore/marchio commerciale
Aven 18023ARS
tecniche
PCR: suitable
Lunghezza
125 mm (4.75 in.)
Descrizione generale
Aven′s innovative Artis Tweezers have set a new precedent for precision. Our cutting edge design features a angled bend at the end of the tip, which provides a softer overall grip with a large surface area that reduces hand fatigue, enhancing productivity.
The durable tweezers are protected by an attractive ESD safe coating, and the highly polished stainless steel tips with rounded edges allow operators to handle very fine wire, miniature parts, electrical components, and other delicate materials in a confined space without risk of damage.
The durable tweezers are protected by an attractive ESD safe coating, and the highly polished stainless steel tips with rounded edges allow operators to handle very fine wire, miniature parts, electrical components, and other delicate materials in a confined space without risk of damage.
Applicazioni
Excellent for general assembly, with strong fine points.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z742669-1EA | 4061835516568 |