Z742593
Accuris™ SmartDoc™
2.0 Imaging System with blue light illumination base, AC/DC input 230 V AC, UK plug
Sinonimo/i:
gel documentation equipment, transilluminator system
About This Item
Prodotti consigliati
Caratteristiche
12V DC power supply included
High Intensity Blue LED; Orange PMMA photo filter and view cover
includes gel prep/cutting platform, blue light illumination base, orange filter cover, imaging enclosure and orange photo filter
Confezionamento
pkg of 1
Produttore/marchio commerciale
Benchmark Scientific E5001-SDB-E -UK
Largh. × prof. × alt.
23 cm × 19 cm × 22 cm, 2.6lbs (1.2kg) weight
Compatibilità
smartphones and tablets with camera
Descrizione generale
Features:
- Ideal for common safe stains (SmartGlow, SYBR Green, GelGreen, etc)
- Visible blue light will not damage DNA
- Quality gel imaging with any smart phone
- Gel access for DNA band cutting
- Imaging enclosure can also be used on existing trans illuminators
- Maximum gel size: 15X15 cm
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z742593-1EA | 4061826162040 |