Z692891
Rota-Rack® Duo tube rack
pkg of 1 ea
About This Item
Codice UNSPSC:
41122804
NACRES:
NB.35
Prodotti consigliati
Materiali
polypropylene
Caratteristiche
Holds 48 x 0.5 mL tubes
Holds 48 x 1.5 mL tubes
Holds 96 x 0.2 mL tubes
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Heathrow Scientific HS29041A
Lungh. × Largh. × Alt.
296 mm × 118 mm × 34 mm
Descrizione generale
Work with PCR tubes right next to centrifuge tubes. The 0.2 mL tube wells are oriented in standard 96-well format to be used with PCR transfer trays.
Rotate the rack to customize for your application. Work with 96 x 0.2 mL tubes and 48 x 0.5 mL tubes or turn a module and work with 96 x 0.2 mL tubes and 48 x 1.5 mL tubes. Flip the rack and work with 48 x 0.5 mL tubes and 48 x 1.5 mL tubes. Or turn a module and work with a total of 96 x 1.5 mL tubes.
Rotate the rack to customize for your application. Work with 96 x 0.2 mL tubes and 48 x 0.5 mL tubes or turn a module and work with 96 x 0.2 mL tubes and 48 x 1.5 mL tubes. Flip the rack and work with 48 x 0.5 mL tubes and 48 x 1.5 mL tubes. Or turn a module and work with a total of 96 x 1.5 mL tubes.
- Tapered wells securely suspend tubes upright and makes tube removal easy
- Imprinted alpha-numeric system makes it easy to quickly identify tubes
- Autoclavable
Note legali
Rota-Rack is a registered trademark of Heathrow Scientific
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)