Documenti fondamentali
Z682683
Sigma-Aldrich
Ace drying flask, pear-shaped, with side indents
capacity 500 mL, joint: ST/NS 29/32, glass
Sinonimo/i:
Aldrich® drying flask, pear-shaped, with side indents
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
glass
pear-shaped flask
single-neck
Giunto
joint: ST/NS29/32
Sterilità
non-sterile
Caratteristiche
closure type Standard Taper Joint
Confezionamento
case of 1
Produttore/marchio commerciale
Büchi equivalent, 00452
Capacità
500 mL
Descrizione generale
These pear shaped drying flasks for rotary evaporators have side indents which increase the mixing action when the flask is rotated on an evaporator .
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)