Z680273
Titanium tweezers
6 piece kit
Sinonimo/i:
Precision Tweezers, Titanium Forceps
About This Item
Codice UNSPSC:
27112105
NACRES:
MA.01
Prodotti consigliati
Produttore/marchio commerciale
Aven
Categorie correlate
Descrizione generale
This set consists of our most popular titanium tweezers in a handy pouch. Tweezers included in the set are Styles 2A, 3c, OO, AA, 5 and 7.
Titanium tweezers do not shed particles like stainless steel tweezers. Titanium is light weight, high strength, non-magnetic material.
Typical application: handling of sensitive components in cleaning/chemical processes at high temperatures, histology, biology, forensics, electron microscopy etc.
- Lightweight (high strength to weight ratio)
- Zero magnetic retentively
- Temperature resistant to 800 °F
- Chemical and corrosion resistant
- Non-solder adhesive
- Bio-compatible (maintain cell integrity, no inflammatory response)
Typical application: handling of sensitive components in cleaning/chemical processes at high temperatures, histology, biology, forensics, electron microscopy etc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z680273-1EA | 4061826711439 |