Z680257
Titanium tweezers
Style 6
About This Item
Codice UNSPSC:
27112105
NACRES:
MA.01
Prodotti consigliati
Produttore/marchio commerciale
Aven
Lunghezza totale
4.75 in. (121 mm)
Categorie correlate
Descrizione generale
Titanium tweezers do not shed particles like stainless steel tweezers. Titanium is light weight, high strength, non-magnetic material.
Typical application: handling of sensitive components in cleaning/chemical processes at high temperatures, histology, biology, forensics, electron microscopy etc.
- Lightweight (high strength to weight ratio)
- Zero magnetic retentively
- Temperature resistant to 800 °F
- Chemical and corrosion resistant
- Non-solder adhesive
- Bio-compatible (maintain cell integrity, no inflammatory response)
Typical application: handling of sensitive components in cleaning/chemical processes at high temperatures, histology, biology, forensics, electron microscopy etc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z680257-1EA | 4061826711415 |