Z677337
Dermatril™ LR nitrile gloves
size L
Sinonimo/i:
disposable glove
About This Item
Codice UNSPSC:
46181504
NACRES:
NB.35
Prodotti consigliati
Materiali
blue gloves
blue nitrile
Misura
L
Sterilità
non-sterile
Caratteristiche
CE compliant
latex free
powder-free
Confezionamento
pack of 50 ea (boxes of 50)
Produttore/marchio commerciale
KCL 742 - 09
KCL 742
Lungh. × spessore
270 mm × 0.11 mm
Descrizione generale
Dermatril LR is an extended cuff, blue nitrile, powder-free glove providing a high level of protection against chemical and bacteriological contamination. The Dermatril LR disposable glove also displays very good physical durability in comparison to conventional disposable gloves.
All Dermatril LR disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
All Dermatril LR disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
- Cleanroom class 100
- Anti-static
- Free from allergenic substances such as thiurames, benzothiazoles and thiourea
- Extended cuff
- Roughened fingertips for better grip
- Rolled cuff edge for easier donning
- CE compliant
- Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 455, EN 61340-5-1, ASTM 1671:2001, US-FED-ST 209E
Note legali
Dermatril is a trademark of KCL GmbH
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z677337-50EA | 4061833069370 |
Z677337-10X50EA | 4061833069363 |