Documenti fondamentali
Z656348
Vacuubrand rotary vane pump oil B, 5L
Pump oil B:high viscosity, low vapour pressure, pack of 5 L
Sinonimo/i:
Vacuubrand 687011
About This Item
Codice UNSPSC:
40151700
NACRES:
NB.79
Confezionamento
pack of 5 L
Produttore/marchio commerciale
Vacuubrand
Descrizione generale
This vacuum pump oil has excellent viscosity characteristics. Its good chemical resistance, low vapor pressure as well as its better stability when pumping oxidants such as acid and alkaline vapors, makes it superior compared to standard mineral oils. This oil is used for the first filling of RE / RZ / RC series pumps. Oils for rotary vane pumps
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)