Documenti fondamentali
Z645761
Chemical spill bucket
Sinonimo/i:
chemical cleanup, spill cleanup, spill safety
About This Item
Codice UNSPSC:
47131905
NACRES:
NB.35
Produttore/marchio commerciale
Clear-spill CK4
Disponibilità
not available in USA and Canada, ROW
Descrizione generale
Kit designed for small to medium spills, ideal for laboratories. Contains 20 pads, 3 x 8 cm by 1.2 m socks, 2 x 30 cm by 35 cm cushion, bags & ties with instructions supplied in a durable container with a snap close lid. Kit will absorb 31 litres.
PLEASE NOTE: KIT NO LONGER CONTAINS PPE
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 7
1 of 7
Lab-Sorb™ granules absorbs aqueous or mixed aqueous and organic solvent waste
Z688436
Lab-Sorb granules
BRAND® bucket, PE-HD capacity 10 L
BR71774
BRAND® bucket, PE-HD
BRAND® bucket, PP volume 15 L
BR72378
BRAND® bucket, PP
BRAND® bucket, PE-HD capacity 5 L
BR71772
BRAND® bucket, PE-HD
Chemizorb® Powder Absorbent for spilled liquids
Supelco
1.02051
Chemizorb® Powder