Key Documents
Z635472
Swiss Boy lab jack plate
for models 110 and 115, L × W 200 mm × 200 mm
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.35
Produttore/marchio commerciale
Grauer
Lungh. × largh.
200 mm × 200 mm
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Lot/Batch Number
Ci dispiace, ma al momento non ci sono COA disponibili online per questo prodotto.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 10
1 of 10
Micro jack platform size 7.5 cm × 7.5 cm, anodized aluminum
Sigma-Aldrich
Z105856
Micro jack
Swiss Boy lab jacks load ≤90 kg (middle position), platform L × W 240 mm × 320 mm, Model 118
Z635383
Swiss Boy lab jacks
Support jack platform L × W 10 cm × 10 cm
Z565814
Support jack
Support jack platform L × W 20 cm × 20 cm, Maxi-Lift
Z565881
Support jack
Support jack platform L × W 20 cm × 20 cm
Z565830
Support jack
Swiss Boy lab jacks load ≤90 kg (with two plates, middle position), platform L × W 400 mm × 500 mm, Model 118
Z635553
Swiss Boy lab jacks
Support jack platform L × W 30 cm × 30 cm
Z565857
Support jack
Support jack platform L × W 15.2 cm × 15.2 cm
Z692417
Support jack