Documenti fondamentali
Z630012
Sigma-Aldrich
IKA® FK 1 flexible coupling for A11 mill
FK 1 flexible coupling, 1/cs
About This Item
Codice UNSPSC:
41101708
NACRES:
NB.72
Caratteristiche
CE compliant
CSA approved
Confezionamento
case of 1
pack of 1
Produttore/marchio commerciale
IKA 23360.00
Diametro
8 mm , clamping plug
Misura
6-10 mm , clamping range
Descrizione generale
The flexible coupling is required for stirring jobs in glass apparatus. A torque of max. 10 Ncm can be tranfered to KPG-glass stirrers. The FK 1 balances inaccuracies of the set-up.
  • Required for stirring tasks using glass stirring rods
  • The flexible coupling compensates for any structural variances
Note legali
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)