Z567612
Aldrich® swivel Benchclamp™
dual adjustment grippy knobs, 3-prong, size S
Sinonimo/i:
bench clamps
About This Item
Codice UNSPSC:
41122413
NACRES:
NB.35
Prodotti consigliati
Misura
S
Capacità ganasce
0-40 mm
Largh. rebbi
6 mm
Descrizione generale
Aldrich benchclamp clamps have a nickel-plated finish and are corrosion resistant. The clamps come with slip-on fiberglass finger covers. The Grippy Knob design has a round nut which is easy to turn making tightening the clamps around glassware fast and convenient. This swivel head allows for the clamp to have any angle of adjustment.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchclamp is a trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z567612-1EA | 4061837637070 |