Z567302
Ace round-bottom pressure flask with thermowell
with Ace-Thred 25 PTFE front-seal plug, capacity 250 mL
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass
round bottom flask
single-neck
Sterilità
non-sterile
Caratteristiche
closure type PTFE plug seal
Confezionamento
pack of 1
Produttore/marchio commerciale
Ace Glass 8417-15
Capacità
250 mL
Diametro
82 mm
Categorie correlate
Descrizione generale
Heavy-wall borosilicate glass flasks are pressure rated to 60 psig at 120°C. The rounded bottom facilitates use in heating mantles and for other round-bottom flask applications. Flasks are supplied complete with #15 Ace-Thred PTFE front seal bushing and FETFE® O-ring.
Replacement FETFE O-rings:
Ace-Thred #Front SealBack Seal
7 Z504661 Z504742
15 Z504858 Z505021
25 Z505056 Z505110
CAUTION: This glassware is not guaranteed against breakage caused by pressure or vacuum. These flasks should never be used if scratched or otherwise damaged.
Replacement FETFE O-rings:
Ace-Thred #Front SealBack Seal
7 Z504661 Z504742
15 Z504858 Z505021
25 Z505056 Z505110
CAUTION: This glassware is not guaranteed against breakage caused by pressure or vacuum. These flasks should never be used if scratched or otherwise damaged.
Note legali
FETFE is a registered trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z567302-1EA | 4061832351193 |