Z562998
Plas-Labs compact glove box
AC/DC input 120 V AC, Plas-Labs 830-ABB
Sinonimo/i:
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
About This Item
Codice UNSPSC:
41103406
Prodotti consigliati
Produttore/marchio commerciale
Plas-Labs 830-ABB
Largh. x prof. × alt. interne
35 in. × 29 in. × 30 in.
Largh. x prof. × alt. esterne
49 in. × 30 in. × 31 in.
Categorie correlate
Descrizione generale
The Plas-Labs compact glove box features cleanliness, safety, complete containment, porability, and a draft-free atmosphere. The tall profile has a small footprint and is especially suited for hazardous materials to be weighed.
The compact glove box chamber is ideal for determining the dry weight or moisture content of aqueous solutions, adhesives, cereals, toxic chemicals, pharmaceuticals, paper, plastics, and light radioactive materials. It minimizes product degradation caused by moisture absorption.
The compact glove box may also be used with analytical balances, top loaders, bench scales and moisture analyzers, and is suitable for handling toxic materials and pharmaceuticals.
The compact glove box chamber is ideal for determining the dry weight or moisture content of aqueous solutions, adhesives, cereals, toxic chemicals, pharmaceuticals, paper, plastics, and light radioactive materials. It minimizes product degradation caused by moisture absorption.
The compact glove box may also be used with analytical balances, top loaders, bench scales and moisture analyzers, and is suitable for handling toxic materials and pharmaceuticals.
- Removable top section for easy installation of equipment
- Adjustable SS clamp
- Ambidextrous Hypalon® gloves with SS O-rings
- Transparent transfer chamber (12 L × 11 in. I.D.) with adjustable vacuum gauge
- White levelling tray for transferring liquids through the transfer chamber
- Four purge valves, two on transfer chamber, two on main chamber
- HEPA and organic vapour canister filters (replaceable)
- Multiple electrical outlet strip, CE UL and CSA approved
Note legali
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)