Key Documents
Z530409
Sigma-Aldrich
Aldrich® short-path distillation head
Joints: ST/NS 29/32 (2), thermometer joint: ST/NS 10/30
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.43
Giunto
thermometer joint: ST/NS10/30
Joints: ST/NS29/32 (2)
Prof. immersione
75 mm
Descrizione generale
Vigreux indents prevent bumping of fine or lightweight column packing into the condenser or fraction cutter when column is flooded.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)