Documenti fondamentali
Z405566
Sigma-Aldrich
J. Young high-vacuum PTFE valve, straight through
stopcock size 6 mm, side arm O.D. 9 mm
About This Item
Codice UNSPSC:
40141600
NACRES:
NB.79
Produttore/marchio commerciale
J. Young
D.E. braccio laterale
9 mm
Misura rubinetto
6 mm
Descrizione generale
The J. Young high-vacuum PTFE valve helps in controlling the atmosphere when using air-sensitive materials.
Features:
  • PTFE piston with unique PTFE-sheathed elastomer O-ring seals
  • Grease-free glass-to-PTFE seals
  • High-vacuum to 10-6 torr
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)