Z303194
Quickfit® Liebig condenser
L 400 mm, joint: ST/NS 19/26
Sinonimo/i:
C1/22/SC
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Giunto
joint: ST/NS19/26
Produttore/marchio commerciale
Quickfit C1/22/SC
Disponibilità
not available in USA and Canada
not available in USA and Canada
Lunghezza
400 mm
Area superficiale
200 cm2
Descrizione generale
Designed to give a turbulent flow in the water jacket for efficient cooling at a comparatively slow rate of water flow. These condensers are adequate and effective in many applications.
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z303194-1EA | 4061826686744 |