Z286494
Waste solvent funnels
for container cap. 8 L, closure size 53B
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.43
Prodotti consigliati
Diam. imbuto
8 in.
Descrizione generale
ECO Funnels prevent waste solvent evaporation. We recommend that the ECO Funnel be removed from the waste container at the end of each day or when it is not in use for long periods of time.
Specifications:
Funnels are sized to fit the following waste solvent containers:
Order funnel and waste container separately.
Specifications:
- Chemically resistant HDPE and PP construction
- Lip-seal on lid (red) seals the waste system when closed
- Funnel stem sealed to waste container cap prevents emission from sides of stem
- Pressure-equalizing tube prevents solvent trapping
Funnels are sized to fit the following waste solvent containers:
- 4 L funnel fits round bottle, B8410
- 8 L funnel fits round bottle, B8535
- 9 L funnel fits rectangular carboy, C6317
- 20 L funnel fits rectangular D.O.T. carboy, Z268208
Order funnel and waste container separately.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z286494-1EA | 4061833495643 |