Z284254
Spinbar® Pyrex® magnetic stir bars
size 1/2 in. × 3/8 in.
Sinonimo/i:
Stirbar, Stirring Bar, glass Stir Bar
About This Item
Codice UNSPSC:
41103806
NACRES:
NB.72
Prodotti consigliati
Materiali
cylindrical
round glass coated
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F37101-0012
Lunghezza
12.7 mm
Diametro
9.5 mm
Misura
1/2 in. × 3/8 in.
Categorie correlate
Descrizione generale
- Spinbar® Pyrex® magnetic stir bars are manufactured from Type I borosilicate glass.
- Ideal for use in applications requiring high temperature up to 274°C (525°F).
- The smooth Pyrex®glass coating ensures easy cleaning and offers “zero absorption” thus preventing cross-contamination of samples.
- Autoclavable.
Note legali
PYREX is a registered trademark of Corning, Inc.
Spinbar is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z284254-1EA | 4061826277485 |