Documenti fondamentali
Z281751
Sigma-Aldrich
Chem-FLEX transfer line
tube L 60 in.
Sinonimo/i:
Flexible transfer line, Transfer tubing
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.43
Lunghezza
60 in.
Categorie correlate
Descrizione generale
Two 12 gauge SS needles (6 and 18 in.) are connected to the CHEM-FLEX 106 tubing with nylon clamps. Liquids contact only PTFE and SS during transfers. For extra security when using large bore transfer needles, fasten septum to bottle or flask opening with a wrap-it tie.
The nylon clamps are replaceable, see Z224170.
Please note: To see all 58 stainless steel 304 syringe needles with non-coring points click here
Note legali
Chem-Flex is a trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 7
1 of 7
Double-tipped needle gauge 12, L 36 in.
Z185205
Double-tipped needle
Double-tipped needle gauge 16, L 36 in.
Z100900
Double-tipped needle
Double-tipped needle gauge 20, L 24 in.
Z101095
Double-tipped needle
Double-tipped needle deflecting tip, L 24 in., gauge 12
Z185213
Double-tipped needle
Double-ended needle gauge 16, L 24 in.
Z102431
Double-ended needle
Global Trade Item Number
SKUGTIN
Z281751-1EA4061833634219