Z278521
Wilmad® coaxial insert
stem L 32 mm, reference capacity 40 μL, sample capacity 339 μL
Sinonimo/i:
Wilmad nmr tubes, nmr sample tubes, nmr tubes
About This Item
Codice UNSPSC:
41121705
NACRES:
SB.32
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Wilmad 520-3
Lungh. stelo
32 mm
Descrizione generale
Precision, all-glass construction virtually eliminates sidebands. Superior to PTFE inserts in variable-temperature experiments. Upper section of insert fits snugly into the outer NMR tube; 2 mm O.D. insert stem is held straight within 0.013 mm. Use inserts with 5 mm O.D. × 7 in. L thin-wall Precision NMR tubes only. Each insert supplied with a closing cap.
Coaxial inserts are applied in external referencing and locking, studying molecular interactions and determining magnetic susceptibility.
Outer tube must be ordered separately based on magnetic field strength.
Coaxial inserts are applied in external referencing and locking, studying molecular interactions and determining magnetic susceptibility.
Outer tube must be ordered separately based on magnetic field strength.
Note legali
Wilmad is a registered trademark of SP Industries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z278521-1EA | 4061832803111 |