Z261580
Luer-to-Tubing Connector
Micro-Mate® female Luer to hose end for 1/4 in. to 5/16 in. I.D. tubing, 316 stainless steel
Sinonimo/i:
environmental, veterinary
About This Item
Codice UNSPSC:
40183110
NACRES:
NB.03
Prodotti consigliati
Materiali
316 stainless steel
Produttore/marchio commerciale
Popper 6453
Categorie correlate
Descrizione generale
Hardware specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. 316 SS fittings are noncontaminating, easy to clean and resistant to corrosion.
Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
Note legali
Micro-Mate is a registered trademark of Cadence Science
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z261580-1EA | 4061832530970 |