Z247820
Aldrich® short-path distillation apparatus
Jacketed head
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.43
Descrizione generale
Threaded 24/40 joints. 100 mL two-neck flask and 50 mL recovery flask.
Replacement parts:
Z174459 - Standard head
Z247839 - Jacketed head
Z174491 - Receiver flask, 50 mL
Z174483 - Two-neck flask, 100 mL
Z405531 - Two-neck flask, 250 mL
Z174475 - Gas-inlet adapter
Z174467 - Gas-inlet tube
Please note: Purchase thermometer separately (Z252980)
Replacement parts:
Z174459 - Standard head
Z247839 - Jacketed head
Z174491 - Receiver flask, 50 mL
Z174483 - Two-neck flask, 100 mL
Z405531 - Two-neck flask, 250 mL
Z174475 - Gas-inlet adapter
Z174467 - Gas-inlet tube
Please note: Purchase thermometer separately (Z252980)
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Ricambio
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z247820-1EA | 4061832131924 |