Z223646
Aldrich® septum-inlet adapter, female
Screw-thread, joint: ST/NS 29/32
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Prodotti consigliati
Giunto
joint: ST/NS29/32
Descrizione generale
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)